Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin alpha Chain (hCG alpha) Sheep anti-Human, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody
Specifications
Antigen | Chorionic Gonadotropin alpha Chain (hCG alpha) |
---|---|
Concentration | 11.7 mg/mL |
Dilution | ELISA 1:500-1:1000 |
Applications | ELISA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062636
|
Novus Biologicals
NB10062636 |
0.5 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB017501
|
Novus Biologicals
NB100626360.1ML |
0.1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Chorionic Gonadotropin alpha Chain (hCG alpha) Polyclonal antibody specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for ELISA.Specifications
Chorionic Gonadotropin alpha Chain (hCG alpha) | |
ELISA 1:500-1:1000 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 0.09% Sodium Azide | |
CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHA1, HCG, LHA, TSHA | |
CGA | |
IgG | |
IgG purified | |
This product is specific for human chorionic gonadotrophin (hCG), a 30kD hormone produced by placental syncytiotrophoblasts. hCG acts to maintain the corpus luteum until the developing placenta is able to produce the required levels of oestrogen and progesterone. hCG shares a common alpha chain with the pituitary hormones FSH, TSH and LH, but has a unique beta chain. This antibody reacts with intact hCG. |
11.7 mg/mL | |
ELISA | |
Unconjugated | |
Sheep | |
Human | |
P01233 | |
1081 | |
Native hCG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title