Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Antibody (4B8B3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | Chorionic Gonadotropin beta Chain (HCG beta) |
---|---|
Clone | 4B8B3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61789
![]() |
Novus Biologicals
NBP261789 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Western Blot, ELISA.Specifications
Chorionic Gonadotropin beta Chain (HCG beta) | |
1 mg/mL | |
Monoclonal | |
Purified | |
Cancer | |
1082 | |
Purified recombinant fragment of human CGB (AA: 21-165) expressed in E. Coli. | |
Primary | |
17.7 kDa |
4B8B3 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
Unconjugated | |
Mouse | |
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB | |
CGB3 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title