Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chorionic Gonadotropin beta Chain (hCG beta) Rabbit anti-Human, Clone: HCGb/1985R, Novus Biologicals™
Rabbit Monoclonal Antibody
$301.00
Specifications
Antigen | Chorionic Gonadotropin beta Chain (hCG beta) |
---|---|
Clone | HCGb/1985R |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122468
|
Novus Biologicals
NBP30749720UG |
20 μg |
Each of 1 for $301.00
|
|
|||||
Description
Chorionic Gonadotropin beta Chain (hCG beta) Monoclonal specifically detects Chorionic Gonadotropin beta Chain (hCG beta) in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
Chorionic Gonadotropin beta Chain (hCG beta) | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer | |
10 mM PBS with 0.05% BSA | |
1082 | |
IgG | |
Protein A or G purified |
HCGb/1985R | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CGB3choriogonadotropin subunit beta, CGB5, CGB7, CGB8, CG-beta, Chorionic gonadotrophin chain beta, chorionic gonadotropin beta 3 subunit, chorionic gonadotropin beta chain, chorionic gonadotropin beta subunit, chorionic gonadotropin, beta polypeptide, hCGB | |
Recombinant full-length Chorionic Gonadotropin beta Chain (hCG beta) protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title