Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CHREBP Antibody (2D9NB) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$206.50 - $426.00
Specifications
Antigen | CHREBP |
---|---|
Clone | 2D9NB |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31289120), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) |
Applications | Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP244307SS
![]() |
Novus Biologicals
NBP244307SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP244307
![]() |
Novus Biologicals
NBP244307 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
CHREBP Monoclonal specifically detects CHREBP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP).Specifications
CHREBP | |
1.0 mg/mL | |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Cancer, Chromatin Research, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
BHLHD14, CHREBPcarbohydrate-responsive element-binding protein, Class D basic helix-loop-helix protein 14, MIOWS basic-helix-loop-helix leucine zipper protein, MLX interacting protein-like, MLX interactor, MLX-interacting protein-like, MONDOBWilliams-Beuren syndrome chromosomal region 14 protein, WBSCR14Williams-Beuren syndrome chromosome region 14 protein 1, Williams Beuren syndrome chromosome region 14, Williams-Beuren syndrome chromosome region 14 protein 2, WS-bHLHbHLHd14carbohydrate response element binding protein | |
MLXIPL | |
IgG2b κ | |
Protein G purified |
2D9NB | |
Western Blot 1:500 ug/ml, Chromatin Immunoprecipitation reported in scientific literature (PMID 31289120), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Q9NP71-1 | |
51085 | |
Partial recombinant human ChREBP protein between amino acids 600-800. [Uniprot: Q9NP71] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title