Learn More
Wooden Molecular Model Building Kit
SCI_ED
Create three-dimensional representations of molecules.
Manufacturer:  Christensen Educational Materials 9077
Catalog No. S44051
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 1 in. (2.54cm) Springs (35), 1.13 in. (2.86cm) springs (15), 1.5 in. (3.8cm) springs (35), black carbon balls (30), white hydrogen balls (30), red oxygen balls (12), green halogen balls (18), orange nitrogen balls (5), and yellow sulfur balls (5)
Description
Description
  • Gain critical hands-on experience with formulas
  • Construct complex molecules for classes from secondary levels through research universities
  • Affordable, high-quality construction
  • Precision-drilled, 0.79 in. dia. (2cm) balls ensure correctness of bond angles
  • Specially drilled carbon balls allow formation of tetrahedron
  • Replacement parts available; see cat. nos. S440511 to -6
Specifications
Specifications
0.75 in. dia. Balls - 100 total, 30 - black/Carbon (Tetrahedral 4 holes), 5 -Yellow/Sulfur (Tetrahedral 4 holes), 5 -Orange/Nitrogen (Tetrahedral 4 holes), 12 - Red/Oxygen (2 holes), 30 -White/Hydrogen (1 hole), 18 -Green/Halogens (1 hole), Bonds - 85 total, 35 - 1 in. Springs/Carbon -Hydrogen, 35 - 1.5 in. Springs/Aliphatic Carbon, 15 - 1 .12 in. Springs/Aromatic Carbon
Molecular Model, Wooden Building Kit