Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRM2 Mouse anti-Human, Mouse, Clone: 1424CT461.78.60, Abnova™

Mouse monoclonal antibody raised against human CHRM2.

Manufacturer:  Abnova Corporation MAB12363

Catalog No. 89-933-973


Add to cart

Description

Description

The muscarinic cholinergic receptors belong to a larger family of G protein-coupled receptors. The functional diversity of these receptors is defined by the binding of acetylcholine to these receptors and includes cellular responses such as adenylate cyclase inhibition, phosphoinositide degeneration, and potassium channel mediation. Muscarinic receptors influence many effects of acetylcholine in the central and peripheral nervous system. The muscarinic cholinergic receptor 2 is involved in mediation of bradycardia and a decrease in cardiac contractility. Multiple alternatively spliced transcript variants have been described for this gene. [provided by RefSeq]

Specifications

Specifications

CHRM2
1424CT461.78.60
Mouse monoclonal antibody raised against human CHRM2.
In PBS (0.09% sodium azide)
FLJ43243/HM2/MGC120006/MGC120007
Mouse
IgG1, kappa
400 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:25) Immunohistochemistry (1:25) Immunofluorescence (1:25) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Liquid
CHRM2
Recombinant protein corresponding to human CHRM2.
Protein G purification
RUO
Primary
1129
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit