Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA10 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548341

Catalog No. PA548341

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

After binding acetylcholine, the achr responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications

Specifications

CHRNA10
Polyclonal
PBS with 0.09% sodium azide
Q9GZZ6
Neuronal acetylcholine receptor subunit alpha-10, Nicotinic acetylcholine receptor subunit alpha-10, NACHR alpha-10, CHRNA10, NACHRA10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57053
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CHRNA10
Liquid
CHRNA10
KLH conjugated synthetic peptide between 179-206 amino acids from the Central region of human CHRNA10.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit