Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRNA2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124293
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124293 100 μg
1 options

Catalog No. PIPA124293

Supplier: Thermo Scientific PA124293

Rabbit Polyclonal Antibody

PA1-24293 detects CHRNA2 in Human samples. PA1-24293 has been successfully used in Western Blot procedures. PA1-24293 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human CHRNA2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

After binding acetylcholine, the ACHR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHRNA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene CHRNA2
Gene Accession No. Q15822
Gene Symbols CHRNA2
Host Species Rabbit
Immunogen Synthetic peptide region of human CHRNA2.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1135
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.