Learn More
Aviva Systems Biology Corp CHRNB2 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP34970_T100
This is a rabbit polyclonal antibody against CHRNB2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CHRNB2. Alternate names: EFNL3, nAChRB2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-52983