Learn More
CHRNB4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221921AP
Catalog No. 50-173-3763
Add to cart
Description
Description
After binding acetylcholine, the AChR responds by an extensive change in conformation that affects all subunits and leads to opening of an ion-conducting channel across the plasma membrane.
Specifications
Specifications
CHRNB4
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30926, Q8R493
CHRNB4
Rabbit
IgG
150 μL
-20°C
Primary
108015, 1143
Flow Cytometry, Western Blot
Unconjugated
CHRNB4
Liquid
CHRNB4
CHRNB4 Fusion Protein Ag16767
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse