Learn More
Aviva Systems Biology Corp CHRND Antibody - N-terminal region (AVARP13022_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP13022_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHRND. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human CHRND. Alternate names: ACHRD, CMS2A, FCCMS, SCCMSApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62608