Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHRND Mouse anti-Human, Clone: 1H1F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261733

Catalog No. NBP2-61733


Add to cart

Description

Description

CHRND Monoclonal antibody specifically detects CHRND in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

CHRND
1H1F9
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Purified
CHRND
Purified recombinant fragment of human CHRND (AA: extra 22-245) expressed in E. Coli.
58.8 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
acetylcholine receptor subunit delta, ACHRDSCCMS, cholinergic receptor, nicotinic, delta, cholinergic receptor, nicotinic, delta polypeptide, CMS2A, FCCMS
Mouse
IgG1
Protein G purified
GPCR, Hypoxia
Primary
1144
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit