Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Chroma-Luc™ Luciferase Reporter Vectors

Catalog No. p-3336306
Click to view available options
Product Type:
pCBG68-Basic Vector
pCBG68-Control Vector
pCBG99-Basic Vector
pCBG99-Control Vector
pCBR-Basic Vector
pCBR-Control Vector

Chroma-Glo™ Luciferase Assay System and the Chroma-Luc Vectors are used to generate red and green (dual-color) luminescence from a single sample upon addition of a single reagent.

  • Control Vectors That Can Generate Red and Green (Dual-Color) Luminescence in a Single Sample
  • Use as the control reporter vector with Chroma-Luc™ Basic Vectors
  • Engineered genes are based on click beetle luciferase
  • Luciferase expression driven by SV40 promoter and enhancer

Specifications

For Use With (Equipment) Chroma-Glo assay system
Promoter SV40
Product Type pCBR-Basic Vector
Reporter Gene Luciferase
Content And Storage -30°C to -10°C
Quantity 20 μg
For Use With (Application) Cloning regulatory elements of interest to produce red/green (dual color) luminescence
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.