Learn More
ChromaCare™ LC-MS Aqueous Rinse, Probe Wash 4, Thermo Scientific™
Ready-to-use blend of water and 2% acetonitrile (v/v) serves as an aqueous wash solution to clean sample probes and needles on LC instruments.
Catalog No. T001282500
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Suitable for cleaning sample preparation needle and drain lines on LC instrument after sampling
  • Save time by using off-the-shelf blend — premade for convenience
  • Helps to maintain clean sample lines and reduce sample carryover
  • Packaged in 2.5L amber glass bottle
  • Complementary to T001262500
Specifications
Specifications
75-05-8
Clear, colorless liquid
Amber Glass Bottle
Undesignated
Water with 2% Acetonitrile
1.8 - 2.2% (Acetonitrile)
≤1 ppm
2.5L