Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ChromaCare™ LC-MS Instrument Flush Solution, Thermo Scientific™
ChromaCare™ LC-MS Instrument Flush Solution is designed to prepare LC/MS instruments for start-up. By reducing background noise, this solution facilitates instrument installation and preventative maintenance routines.
Specifications
CAS | 75-05-8; 67-56-1; 67-63-0; 7732-18-5 |
---|---|
Color | Colorless |
Percent Purity | 25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) |
Grade | LC-MS |
Physical Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
T111101000
|
Fisher Chemical
T111101000 |
1 L | Borosilicate glass bottle |
Each for $167.70
Case of 6 EA for $798.50
Order in Bulk
Request bulk or custom formats |
N/A | ||||
Description
- Designed by Thermo Scientific to maximize cleaning while preserving instrument integrity in Vanquish™ and UltiMate™ UHPLC systems with QQQ MS
- Comprised of a unique blend of polar protic and aprotic solvents for removing a wide variety of contaminants
- Particularly suitable for new LC/MS and UHPLC/MS installations to rapidly stabilize baseline
- A blend of 25% acetonitrile, 25% methanol, 25%, water, and 25% 2-propanol (IPA)
- Offered in 1L borosilicate bottles and 2.5L amber glass bottles
Specifications
75-05-8; 67-56-1; 67-63-0; 7732-18-5 | |
25% Acetonitrile, 25% Methanol, 25% Water, 25% 2-Propanol (IPA) | |
Liquid |
Colorless | |
LC-MS |
RUO – Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title