Thermo Scientific Chromacol 9mm Screw Cas Available on GSA/VA Contract for Federal Government customers only.

Thermo Scientific™ Chromacol 9mm Screw Caps are offered with a selection of liner materials and colors.

Manufacturer: Thermo Scientific 9SCBST1X

 View more versions of this product

Catalog No. 14-824-307

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Part Not Found  Please contact customer service at 1-800-766-7000 with error code:999. 


    Specifications

    Description 9mm Screw Cap Prefitted Silicone/PTFE Liner Pre-cut; 6mm opening; Blue
    Closure Color Blue
    Closure Material Prefitted Silicone/PTFE Liner Pre-cut
    Pre-slit Yes
    Vial Closure Size 9mm
    Type Open Top Screw Cap
    Item Description 9-SC(B)-ST1X
    Closure Liner Prefitted Silicone/PTFE Liner

    Thermo Scientific™ Chromacol 9mm Screw Caps are offered with a selection of liner materials and colors.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit