Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Chromacol 9mm Screw Cas
Thermo Scientific™ Chromacol 9mm Screw Caps are offered with a selection of liner materials and colors.
Specifications
Closure Color | Black |
---|---|
Pre-slit | No |
Product Line | Chromacol |
Closure Size | 9 mm |
Catalog Number | Mfr. No. | Closure Color | Pre-slit | Liner Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Color | Pre-slit | Liner Material | Price | Quantity & Availability | |||
14-823-622
|
Thermo Scientific™
9SCJST101 |
Black | No | Prefitted Soft Silicone/PTFE | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Thermo Scientific™ Chromacol 9mm Screw Caps are offered with a selection of liner materials and colors.
Specifications
Black | |
No | |
Chromacol | |
9 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title