Thermo Scientific Chromacol Select Kits for Agilent™ Instruments

Thermo Scientific™ Chromacol Select Kits for Agilent Instruments have contents carefully chosen to match the needs of these popular autosamplers.

Manufacturer: Thermo Scientific HPLS

 View more versions of this product

Catalog No. 03-341-918

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With Agilent 1100
    Type Assembled Vial Kit
    Vial Closure Size 9mm
    Vial Volume (Metric) 2mL
    Closure Type Screw Top
    Item Description Agilent 1100, 2mL Screw Top Vial
    Description 2mL Screw Top Vial with a Type 8 Rubber/PTFE Seal (Agilent 1100)
    Septa Type 8 Rubber/PTFE

    Thermo Scientific™ Chromacol Select Kits for Agilent Instruments have contents carefully chosen to match the needs of these popular autosamplers.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit