Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Chroman-8-ylmethyl)amine, 90%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
933727-40-3
C10H13NO
163.22
MFCD17171385
CPJFACCHHWIMEM-UHFFFAOYSA-N
chroman-8-ylmethyl amine,2h-1-benzopyran-8-methanamine, 3,4-dihydro,1-3,4-dihydro-2h-1-benzopyran-8-yl methanamine
82596485
3,4-dihydro-2H-chromen-8-ylmethanamine
C1CC2=C(C(=CC=C2)CN)OC1
933727-40-3
C10H13NO
163.22
MFCD17171385
CPJFACCHHWIMEM-UHFFFAOYSA-N
chroman-8-ylmethyl amine,2h-1-benzopyran-8-methanamine, 3,4-dihydro,1-3,4-dihydro-2h-1-benzopyran-8-yl methanamine
82596485
3,4-dihydro-2H-chromen-8-ylmethanamine
C1CC2=C(C(=CC=C2)CN)OC1
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Formula Weight 163.22
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material (Chroman-8-ylmethyl)amine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger