Chromate, Ion Chromatography Standard Solution, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 42564AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42564AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula K2CrO4 in H2O
    MDL Number MFCD00011368
    Chemical Name or Material Chromate, CrO4?2 1000μg/mL

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 500mL
    EINECS Number 232-140-5
    Odor Odorless
    MDL Number MFCD00011368
    Health Hazard 1 H340-H350: May cause genetic defects. May cause cancer.
    Molecular Formula K2CrO4 in H2O
    Appearance Clear liquid
    Chemical Name or Material Chromate, CrO4?2 1000μg/mL

    H340-H350: May cause genetic defects. May cause cancer.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit