Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahlstrom-Munksjo Chromatography, Electrophoresis and Blotting Paper, Grade 222

Offers extremely high flow and is recommended for both electrophoresis of large molecules and as a blotter to complete the blotting sandwich in gel transfer applications.

$59.40 - $283.50

Specifications

Flow Rate High
For Use With (Application) Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
Grade 222
Thickness (Metric) 0.83 mm
View More Specs

Products 4
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
09-790-48
View Documents
Ahlstrom-Munksjo
22288105
80 x 105 mm
Pack of 100 for $59.40
Add to Cart
 
09-790-26B
View Documents
Ahlstrom-Munksjo
22203545SPECIAL
350 x 450 mm
Pack of 25 for $139.60
Add to Cart
 
09-790-49J
View Documents
Ahlstrom-Munksjo
22282020
200 x 200 mm
Pack of 100 for $283.50
Add to Cart
 
09-790-49F
View Documents
Ahlstrom-Munksjo
22281416
140 x 160 mm
Pack of 100 for $205.00
Add to Cart
 
Description

Description

Specifications

Specifications

High
Electrophoresis of large molecules and as blotter to complete the blotting sandwich in gel transfer
222
0.83 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.