Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Chromatography Filter Paper
SCI_ED

Chromatography filter paper sheet.

Supplier:  Eisco™ CH0395C

Catalog No. S23810


Add to Cart

Description

Description

  • Chromatography filter paper sheet.
  • Size of 25 x 25 cm.
Specifications

Specifications

25 cm
1 in.
1 in.
Chromatography Filter Paper
Chromatography
Chromatography
Paper
25 cm
25 x 25 cm
Sheet
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit