Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Chromatography Funnels

For use with Solvent Addition in Flash Chromatography

Supplier:  MilliporeSigma™ Supelco™ Z4007261EA

 View more versions of this product

Catalog No. 11-101-1847


Description

Description

  • Use for solvent addition in flash chromatography
  • Designed to not disturb the top layer of the column
  • Perforated stem with a closed-end allows solvent to pour down column walls
TRUSTED_SUSTAINABILITY
Specifications

Specifications

108 mm
Polypropylene
Solvent Addition in Flash Chromatography
Chromatography Funnel
240 mL
115 mm
38 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.