Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ ChromatoPur™ Bovine IgG, ImmunoAssay
Bovine IgG is suitable as a standard and coating reagent for ELISA
$144.00 - $3493.71
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP08641403
|
MP Biomedicals™
08641403 |
100 g |
Each for $741.00
|
|
ICN641401
|
MP Biomedicals™
08641401 |
1 g |
Each for $144.00
|
|
MP8641404
|
MP Biomedicals™
08641404 |
1 kg |
Each for $3,493.71
|
|
Description
Purified using gentle chromatographic extraction.
Physical Characteristics
- Appearance: Off-white powder
- Purity (SDS-PAGE): ≥95% (IgG w/w Total Protein
- Total Protein (anhydrouse): ≥96%
- pH (4%): 4.0 to 5.0
- Moisture: ≤10%
- Sodium: ≤1.5% w/w
- Chloride: ≤2.4% w/w
Certifications: ISO, cGMP, EDQM
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title