Learn More
Supply SolutionsSupplier Diversity Partner Hach - Chromium Reagent, Chromaver 3, Hexavalent, 5Ml Or 10Ml Sample, Powder Pillow. Hach Chemicals. For Determination Of Hexavalent Chromium By The Diphenylcarbohydrazide Method DFS Item
Manufacturer:  Supply SolutionsSupplier Diversity Partner HACH 1271099
For determination of hexavalent chromium by the Diphenylcarbohydrazide method. USEPA accepted for reporting for wastewater analyses using Hach Method 8023, Diphenylcarbohydrazide Method. Photometric Sample Size: 10ml. Photometric Range: up to 0.70 mgL. Kit Sample Size: 5ml. Kit Range: Up to 1.50 Pack of 100 powder pillows. > To confirm accurate results, see appropriate standard solutions on the Consumables tab.; Range: 0.010 to 0.700 mgL Cr6+; Instrument: All except DR3000, DR2000, DR 900; Platform : Powder Pillows; Number of tests: 100; Method: 8023; EPA compliant: Yes
Catalog No. 50-476-296