Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Acid, 10% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
1333-82-0,7732-18-5
1333-82-0,7732-18-5
Quantity 4 L
Grade Chemical Solution
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.