Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromic Chloride, Hexahydrate, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
10060-12-5
Cl3CrH12O6
266.44
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
10060-12-5
Cl3CrH12O6
266.44
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium(3+) hexahydrate trichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

C1344, 10060-12-5, CrCl3•6H2O

Spectrum™ Chromic Chloride, Hexahydrate, USP is intended for use as an additive to intravenous solutions for total parenteral nutrition. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Insoluble Matter 0.01%
Percent Purity 98-101%
Packaging Fiber Drum
Quantity 50 kg