Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-4781859
Click to view available options
Quantity:
100 mL
500 mL
7789-02-8, 7697-37-2, 7732-18-5
CrN3O9
238.01
MFCD00010944
PHFQLYPOURZARY-UHFFFAOYSA-N
9887206
chromium(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
7789-02-8, 7697-37-2, 7732-18-5
CrN3O9
238.01
MFCD00010944
PHFQLYPOURZARY-UHFFFAOYSA-N
9887206
chromium(3+);trinitrate;nonahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mL
Color Colorless
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5%; Chromium(III) nitrate nonahydrate: 0.77%; Water: 94.23%
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Chemical Name or Material Chromium, AAS Standard Solution
Hazard Category H314-H332-H335
Hazard Statement When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit