Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Chloride, X-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

10060-12-5
Cl3CrH12O6
266.436
LJAOOBNHPFKCDR-UHFFFAOYSA-K
104957
CHEBI:53442

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
SMILES O.O.O.O.O.O.Cl[Cr](Cl)Cl
IUPAC Name trichlorochromium;hexahydrate
Formula Weight 266.45
Packaging Poly Bottle
Specific Gravity 1L = 1.76kg
Quantity 500g
Chemical Name or Material Chromium Chloride, X-Hydrate
Grade BAKER ANALYZED™ Reagent