Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Chloride, X-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
10060-12-5
Cl3CrH12O6
266.436
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate,chromic chloride hexahydrate,hexaaquachromium chloride,chromium trichloride hexahydrate,trichlorochromium hexahydrate,chromium chloride hexahydrate,hexaaquachromium iii chloride,chrom iii-chlorid,chromium 3+ chloride hexahydrate,chlorid chromity hexahydrat czech
104957
CHEBI:53442
trichlorochromium;hexahydrate
O.O.O.O.O.O.Cl[Cr](Cl)Cl
10060-12-5
Cl3CrH12O6
266.436
LJAOOBNHPFKCDR-UHFFFAOYSA-K
chromium iii chloride hexahydrate,chromic chloride hexahydrate,hexaaquachromium chloride,chromium trichloride hexahydrate,trichlorochromium hexahydrate,chromium chloride hexahydrate,hexaaquachromium iii chloride,chrom iii-chlorid,chromium 3+ chloride hexahydrate,chlorid chromity hexahydrat czech
104957
CHEBI:53442
trichlorochromium;hexahydrate
O.O.O.O.O.O.Cl[Cr](Cl)Cl

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 266.45
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.76kg
Quantity 2.5kg
Chemical Name or Material Chromium Chloride, X-Hydrate