Alfa Aesar™ Chromium(III) chloride hexahydrate, 98%

Manufacturer: Alfa Aesar™ 42113A1

Quantity 1kg

 View more versions of this product

Catalog No. AA42113A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10060-12-5
    Formula Weight 266.45 (158.35 Anhydrous)
    MDL Number MFCD00149660
    UN Number UN3260
    Merck Index 14,2222
    Chemical Name or Material Chromium(III) chloride hexahydrate

    Specifications

    Formula Weight 266.45 (158.35 Anhydrous)
    Health Hazard 2 GHS H Statement
    H334-H314-H318-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,2222
    Quantity 1kg
    RTECS Number GB5450000
    Odor Odorless
    MDL Number MFCD00149660
    UN Number UN3260
    Solubility Information Slightly soluble in acetone; Soluble in water, alcohol; Insoluble in ether
    TSCA TSCA
    Physical Form Crystalline
    CAS 10060-12-5
    Melting Point 83°C
    Health Hazard 1 Danger
    Density 1.835g/mL
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Color Dark Green
    Chemical Name or Material Chromium(III) chloride hexahydrate
    Assay 98%

    GHS H Statement
    H334-H314-H318-H302-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit