Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium(III) chloride hexahydrate, 98%, ACROS Organics™
Click to view available options
Quantity:
500g
5kg
Packaging:
Plastic bottle
Plastic drum
Specifications
Assay Percent Range | 98% |
Linear Formula | [Cr(H2O)4Cl2]Cl·2H2O |
Merck Index | 14, 2222 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in acetone,practically insoluble in ether |
Formula Weight | 266.44 |
Physical Form | Crystalline Powder |
Percent Purity | 98% |
Iron (Fe) | 300ppm max. |
Packaging | Plastic bottle |
Solubility | Solubility in water: soluble. Other solubilities: soluble in alcohol, slightly soluble in acetone, practically insoluble in ether |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK
GHS Signal Word: Warning