Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Acros Organics

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
10170-68-0
C12H24Cl3CrO3
374.67
MFCD00013222
CYOMBOLDXZUMBU-UHFFFAOYSA-K
chromium iii chloride tetrahydrofuran complex 1:3,chromium iii chloride tetrahydrofuran adduct,crcl3 thf 3,crcl3 3thf,chromium iii chloride tetrahydrofuran complex,chromium chloride; tris tetrahydrofuran,chromium iii chloride complex 1:3 tetrahydrofuran
11100829
oxolane;trichlorochromium
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
10170-68-0
C12H24Cl3CrO3
374.67
MFCD00013222
CYOMBOLDXZUMBU-UHFFFAOYSA-K
chromium iii chloride tetrahydrofuran complex 1:3,chromium iii chloride tetrahydrofuran adduct,crcl3 thf 3,crcl3 3thf,chromium iii chloride tetrahydrofuran complex,chromium chloride; tris tetrahydrofuran,chromium iii chloride complex 1:3 tetrahydrofuran
11100829
oxolane;trichlorochromium
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
Assay Percent Range 98%
Formula Weight 374.67
Physical Form Crystalline Powder
Percent Purity 98%
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Color Purple
Melting Point 143.0°C
Quantity 1g
Chemical Name or Material Chromium(III) chloride tetrahydrofuran complex (1:3)
Show More Show Less

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.

GHS P Statement:
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning