Alfa Aesar™ Chromium(III) fluoride hydrate

Manufacturer: Alfa Aesar™ 1233930

Quantity 250g

 View more versions of this product

Catalog No. AA1233930

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123333-98-2
    Formula Weight 108.99 (Anhydrous)
    MDL Number MFCD00016045
    UN Number UN1756
    Merck Index 14,2223
    Chemical Name or Material Chromium(III) fluoride hydrate

    Specifications

    Formula Weight 108.99 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00016045
    Melting Point >1000°C
    Physical Form Crystalline
    Sensitivity Hygroscopic
    UN Number UN1756
    Color Green
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CHROMIC FLUORIDE, SOLID
    TSCA TSCA
    CAS 123333-98-2
    Merck Index 14,2223
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Chemical Name or Material Chromium(III) fluoride hydrate

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit