Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chromium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1300506

Quantity 5g

 View more versions of this product

Catalog No. AA1300506

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

20249-21-2
238.01 (Anhydrous)
UN2720
Chromium(III) nitrate hydrate
Cr(NO3)3·xH2O
MFCD00010946
14,2226
Puratronic™

Specifications

20249-21-2
Cr(NO3)3·xH2O
MFCD00010946
14,2226
60°C
Chromium(III) nitrate hydrate
Puratronic™
Crystalline
238.01 (Anhydrous)
UN2720
Dark purple
5g
99.999%, (metals basis)

Safety and Handling

GHS H Statement
H272-H301-H332-H319
May intensify fire
oxidizer.
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P221-P210-P301+P310-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CHROMIUM NITRATE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit