Chromium(III) oxide, 99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1228522

Quantity 100g

 View more versions of this product

Catalog No. AA1228522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1308-38-9
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula Cr2O3
    Formula Weight 151.99
    MDL Number MFCD00010949
    Chemical Name or Material Chromium(III) oxide

    Specifications

    Formula Weight 151.99
    Quantity 100g
    Color Pale to dark green
    Density 5.2g/cm3at 20°C
    EINECS Number 215-160-9
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99%
    Molecular Formula Cr2O3
    Appearance Pale to dark green powder
    MDL Number MFCD00010949
    CAS 1308-38-9
    Health Hazard 1 H302-H332-H317 : Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction.
    Chemical Name or Material Chromium(III) oxide

    H302-H332-H317 : Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit