Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 41058KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41058KS


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1308-38-9
151.99
MFCD00010949
Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick

Specifications

1308-38-9
151.99
5.22g/mL
3000°C
2439°C
Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Target
MFCD00010949
Odorless
Pale to dark green
1 each
99.8%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GB6475000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit