Alfa Aesar™ Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.8% (metals basis)

Manufacturer: Alfa Aesar™ 41059KS

Quantity 1 each

 View more versions of this product

Catalog No. AA41059KS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1308-38-9
    Formula Weight 151.99
    MDL Number MFCD00010949
    Chemical Name or Material Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick

    Specifications

    Formula Weight 151.99
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1 each
    Health Hazard 2 GHS H Statement
    H302-H332-H317
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.
    Density 5.22g/mL
    RTECS Number GB6475000
    Odor Odorless
    MDL Number MFCD00010949
    Boiling Point 3000°C
    Melting Point 2435°C
    Health Hazard 1 Warning
    Physical Form Target
    Color Pale to dark green
    TSCA TSCA
    CAS 1308-38-9
    Chemical Name or Material Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
    Assay 99.8%, (metals basis)

    GHS H Statement
    H302-H332-H317
    Harmful if swallowed.
    Harmful if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit