Learn More
Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Description
It is used for tanning of leather and as a photographic film hardener. It is also used in textile dye, photography, ceramics, and the preparation of chromium standard solutions, as a catalyst, mordant, ceramics, tannage and developer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 89°C |
Color | Violet |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 99.9% (Metal basis) |
Solubility Information | Soluble in water 24g/100 mL (15°C). |
Formula Weight | 499.39 (283.21 Anhydrous) |
Odor | Odorless |
Appearance | Violet crystals |
Density | 1.83 g/cm3 |
Show More |
Safety and Handling
Hazard Category | H315-H319 |
Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
EINECSNumber | 233-401-6 |
RTECSNumber | GB6850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.