Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium(III) standard solution, for AAS, 1 mg/ml Cr in 2% HCl, ACROS Organics™

Click to view available options
Quantity:
100mL
For atomic adsorption spectroscopy
CAS 7440-47-3,7647-01-0,7732-18-5
Molecular Formula Cr
MDL Number MFCD00143970
Concentration 997 to 1003 μg/mL of Cr(3+) in 2% HCl (20°C)
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES [Cr]
IUPAC Name chromium
Molecular Weight (g/mol) 51.996
PubChem CID 23976
ChEBI CHEBI:28073
Formula Weight 52
Physical Form Solution
Grade AAS
Packaging Plastic bottle
Density 0.9800g/mL
Color Blue-Green
Quantity 100mL
Chemical Name or Material Chromium(III) standard solution, 1 mg/ml Cr in 2% HCl
Show More Show Less

GHS H Statement:
May be corrosive to metals.

GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning