Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Metal, 2 in. and Finer, Chips, 99%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
7440-47-3
7440-47-3
Quantity 500 g
CAS Max % 1
Packaging Poly Bottle
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.