Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Nitrate, Crystal, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7789-02-8
7789-02-8
Grade Reagent
CAS Max % 1
Percent Purity 0.99
Chloride 0.0005
Insoluble Matter 0.0001
Iron (Fe) 0.0002
Packaging Amber Glass Bottle
Sulfate 0.0001
Quantity 25 g

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.