Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Oxide, Powder, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle

Chemical Identifiers

1308-38-9
Cr2O3
151.989
QDOXWKRWXJOMAK-UHFFFAOYSA-N
517277
CHEBI:48242
oxo(oxochromiooxy)chromium
O=[Cr]O[Cr]=O

A grade of chemicals for laboratory use

Formula Weight 151.99
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 5.10kg
Quantity 2.5kg
Chemical Name or Material Chromium Oxide, Formerly C.P.