Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chromium pieces, 2-3mm (0.08-0.12 in.) thick, 99.995% (metals basis)

Click to view available options
Quantity:
50g
250g
7440-47-3
Cr
51.996
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159
23976
CHEBI:28073
chromium
[Cr]
7440-47-3
Cr
51.996
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom,chrome,cromo,metal,chrome french,compounds,chrom german,chromium, elemental,chromium, metal,ccris 159
23976
CHEBI:28073
chromium
[Cr]
Quantity 50g
Odor Odorless
Physical Form Broken Plate, Large Irregular Pieces
Percent Purity 99.995%
Chemical Name or Material Chromium pieces, 2 to 3mm (0.08 to 0.12 in.) thick
Assay (metals basis)

EINECSNumber : 231-157-5

TSCA : Yes

Recommended Storage : Ambient temperatures