Chromium Powder, -100 mesh, 99% (Metals basis), Alfa Aesar™

Apparent density 2.0-3.0g/cm3

Manufacturer: Alfa Aesar 1014709

Quantity 10g

 View more versions of this product

Catalog No. AA1014709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-47-3
    Physical Form Powder
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00010944
    Chemical Name or Material Chromium

    Specifications

    Quantity 10g
    Color Gray
    DOT Information DOT Class: 4.1, Packaging Group: III
    EINECS Number 231-157-5
    Physical Form Powder
    Assay Percent Range 99% (Metals basis)
    MDL Number MFCD00010944
    Odor Odorless
    Health Hazard 1 H228-H351; Flammable solid. Suspected of causing cancer.
    CAS 7440-47-3
    Appearance Gray powder
    Health Hazard 2 Wear suitable protective clothing and gloves.
    Chemical Name or Material Chromium

    Wear suitable protective clothing and gloves.

    H228-H351; Flammable solid. Suspected of causing cancer.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit