Chromium powder, -200 mesh, 99.95% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1379614

Quantity 25g

 View more versions of this product

Catalog No. AA1379614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-47-3
    MDL Number MFCD00010944
    UN Number UN3089
    Chemical Name or Material Chromium powder

    Specifications

    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00010944
    CAS 7440-47-3
    UN Number UN3089
    Mesh Size −200 mesh
    Odor Odorless
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228-H351;
    Flammable solid.
    Suspected of causing cancer.
    Health Hazard 3 GHS P Statement
    P210-P280-P281-P308+P313-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Silver
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
    RTECS Number GB4200000
    Chemical Name or Material Chromium powder
    Assay 99.95%, (metals basis)

    GHS H Statement
    H228-H351;
    Flammable solid.
    Suspected of causing cancer.

    GHS P Statement
    P210-P280-P281-P308+P313-P405-P501a;
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit