Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium powder, 99.99% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]
7440-47-3
Cr
52.00
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
23976
CHEBI:28073
chromium
[Cr]
Quantity 5g
Appearance Gray powder
Assay Percent Range 99.996% (Metals basis)
Mesh Size -10+20 Mesh
Odor Odorless
Physical Form Powder
Vapor Pressure 0 hPa
Color Gray
Chemical Name or Material Chromium

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-157-5

TSCA : Yes

Recommended Storage : Ambient temperatures