Learn More
Chromium selenide, Black powder, 99.5% (Metals basis), Thermo Scientific Chemicals
Description
It is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications. It is used for coating solar energy or fuel cells and flip-chip applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.5% (Metals basis) |
Formula Weight | 130.96 |
Odor | Odorless |
Density | 6.74g/cm3 |
Chemical Name or Material | Chromium selenide |
Safety and Handling
Hazard Category | H301-H331-H373-H400-H410; Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. |
Hazard Statement | When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. |
DOTInformation | DOT Class: 6.1, Packaging Group: II |
EINECSNumber | 234-999-1 |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.