Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chromium silicide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)
Click to view available options
Quantity:
1 ea.
Specifications
MDL Number | MFCD00168070 |
Formula Weight | 108.17 |
Percent Purity | 99.5% |
Quantity | 1 ea. |
Chemical Name or Material | Chromium silicide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick |
Assay | (metals basis) |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
EINECSNumber : 234-633-0
TSCA : Yes
Recommended Storage : Ambient temperatures