Alfa Aesar™ Chromium silicon monoxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 3566709

Quantity 10g

 View more versions of this product

Catalog No. AA3566709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37264-20-3
    MDL Number MFCD02091702
    Chemical Name or Material Chromium silicon monoxide

    Specifications

    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Mesh Size −8+20 Mesh
    MDL Number MFCD02091702
    CAS 37264-20-3
    Quantity 10g
    Health Hazard 1 Danger
    Avg. Mol. Wt. or Mol. Wt. Range Cr:SiO; 50:50 wt%
    Health Hazard 2 GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.
    Chemical Name or Material Chromium silicon monoxide
    Assay 99.9%, (metals basis)

    GHS H Statement
    H334-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P342+P311-P363-P501a
    Wear respiratory protection.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
    Wash contaminated clothing before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit